Learn More
Research Products International Corp Laboratory Balance, 5 x 5 3/4 Inch Platform, 0.1 Gram Readability, 1500 Gram Capacity
SDP
Catalog No. 50212242
Encompass
Quality construction provides reliable and accurate results. Features ultra-fast micro-processor for enhanced performance and exceptional stability. External 'one button' calibration.
Applications include: counting; percent weighing; totaling; display hold; specific gravity; 14 mass unit conversions; parts counting; look down capability and more.
Catalog No. 50-212-242 Supplier Research Products International Corp Supplier No. 141321
Quality construction provides reliable and accurate results. Features ultra-fast micro-processor for enhanced performance and exceptional stability. External 'one button' calibration.
Applications include: counting; percent weighing; totaling; display hold; specific gravity; 14 mass unit conversions; parts counting; look down capability and more.