Learn More
Research Products International Corp Laboratory Balance, 410 Gram Capacity, 0.01 Gram Readability, 4 Inch Platform
SDP
Catalog No. 50212240
Encompass
Quality construction provides reliable and accurate results. Features ultra-fast micro-processor for enhanced performance and exceptional stability. External 'one button' calibration.
Applications include: counting; percent weighing; totaling; display hold; specific gravity; 14 mass unit conversions; parts counting; look down capability and more.
Catalog No. 50-212-240 Supplier Research Products International Corp Supplier No. 141319
Quality construction provides reliable and accurate results. Features ultra-fast micro-processor for enhanced performance and exceptional stability. External 'one button' calibration.
Applications include: counting; percent weighing; totaling; display hold; specific gravity; 14 mass unit conversions; parts counting; look down capability and more.