Learn More
Medchemexpress LLC AAPK-25 | 2247919-28-2 | 98.1% | 442.32 | C21H13Cl2N3O2S | 10MG
SDP
Catalog No. 5000235873
Encompass_Preferred
AAPK-25 is a small-molecule research compound that acts as a dual inhibitor of Aurora kinases (Aurora-A, -B, -C) and Polo-like kinases (PLK-1, -2, -3). It exhibits antiproliferative activity in several cancer cell lines and is supplied as an off-white to light yellow solid for in vitro use. Solubility in DMSO is 50 mg/mL, and recommended storage is 4°C with protection from light.
  • Dual inhibition of Aurora and PLK kinases demonstrated by low nanomolar Kd values.
  • Exhibits antiproliferative activity across multiple cancer cell lines.
  • High purity suitable for in vitro biochemical and cellular assays.
  • Available in multiple solid package sizes to support different study scales.
  • Readily soluble in DMSO for assay formulation.
  • Defined storage and in-solvent stability for consistent results.
Catalog No. 50-002-35873 Supplier Medchemexpress LLC Supplier No. HY12624910MG
AAPK-25 is a small-molecule research compound that acts as a dual inhibitor of Aurora kinases (Aurora-A, -B, -C) and Polo-like kinases (PLK-1, -2, -3). It exhibits antiproliferative activity in several cancer cell lines and is supplied as an off-white to light yellow solid for in vitro use. Solubility in DMSO is 50 mg/mL, and recommended storage is 4°C with protection from light.
  • Dual inhibition of Aurora and PLK kinases demonstrated by low nanomolar Kd values.
  • Exhibits antiproliferative activity across multiple cancer cell lines.
  • High purity suitable for in vitro biochemical and cellular assays.
  • Available in multiple solid package sizes to support different study scales.
  • Readily soluble in DMSO for assay formulation.
  • Defined storage and in-solvent stability for consistent results.