Learn More
Medchemexpress LLC Aapk-25 | 2247919-28-2 | 98.1% | 442.32 | C21H13Cl2N3O2S | 50MG
SDP
Catalog No. 5000239640
Encompass_Preferred
AAPK-25 is a small-molecule dual inhibitor of Aurora kinases and Polo-like kinases (PLK) that induces mitotic delay and prometaphase arrest, making it useful for cell-based and biochemical studies of mitosis and kinase signaling. Supplied as an off-white to light-yellow solid with high reported purity and available in a 50 mg quantity.
  • Dual Aurora and PLK inhibition providing mechanistic relevance for mitotic studies.
  • Demonstrated anti-tumor activity in preclinical assays.
  • High reported purity (98.09%).
  • Available as a 50 mg solid for laboratory experiments.
  • Soluble in DMSO; ultrasonication may be required for complete dissolution.
  • Stock solutions stable at -80°C for up to 6 months and at -20°C for up to 1 month when protected from light.
Catalog No. 50-002-39640 Supplier Medchemexpress LLC Supplier No. HY12624950MG
AAPK-25 is a small-molecule dual inhibitor of Aurora kinases and Polo-like kinases (PLK) that induces mitotic delay and prometaphase arrest, making it useful for cell-based and biochemical studies of mitosis and kinase signaling. Supplied as an off-white to light-yellow solid with high reported purity and available in a 50 mg quantity.
  • Dual Aurora and PLK inhibition providing mechanistic relevance for mitotic studies.
  • Demonstrated anti-tumor activity in preclinical assays.
  • High reported purity (98.09%).
  • Available as a 50 mg solid for laboratory experiments.
  • Soluble in DMSO; ultrasonication may be required for complete dissolution.
  • Stock solutions stable at -80°C for up to 6 months and at -20°C for up to 1 month when protected from light.