Learn More
Aviva Systems Biology Corp AARS Antibody - C-terminal region (ARP40369_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40369_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AARS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human AARS. Alternate names: CMT2NApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57414