Learn More
Aviva Systems Biology Corp AASDH Antibody - middle region (ARP53414_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53414_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AASDH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AASDH. Alternate names: ACSF4, LYS2, NRPS1098, NRPS998ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61019