Learn More
Aviva Systems Biology Corp AATF Antibody - N-terminal region (ARP38957_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38957_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against AATF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AATF. Alternate names: DED, CHE1, CHE-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56864