Learn More
New England Biolabs, Inc. AatII – 2500 units
SDP
Catalog No. 50811954
Encompass
A restriction endonuclease that recognizes the sequence G_ACGT^C.
Catalog No. 50-811-954 Supplier New England Biolabs, Inc. Supplier No. R0117L
A restriction endonuclease that recognizes the sequence G_ACGT^C.