Learn More
Aat Bioquest AATOM™ 647N DBCO
SDP
Catalog No. NC3919079
Encompass_Preferred
AATOM™ 647n DBCO is an azide-reactive, red fluorescent dye with an excitation/emission of 646/666 nm that is used for FACS and FISH applications.
Catalog No. NC3919079 Supplier Aat Bioquest Supplier No. 2837
AATOM™ 647n DBCO is an azide-reactive, red fluorescent dye with an excitation/emission of 646/666 nm that is used for FACS and FISH applications.