Learn More
American Research Products Inc AAV2 Titration ELISA, Quantitative AAV2 Capids Detection, Sandwich ELISA, 96 Tests
Supplier:  American Research Products Inc PRATV
Encompass
The AAV2 Titration ELISA is a highly sensitive assay designed for the quantification of Adeno-Associated Virus Type 2 capsids, making it essential for gene therapy research. It enables reliable titration of both intact and empty capsids, utilizing monoclonal antibodies for accurate detection. This product stands out for its consistent performance and colorimetric readout, ensuring precise quantification critical for advancing therapeutic applications.
Catalog No. NC1473166