Learn More
ACROBiosystems AAV AAV5 96tests
Supplier:  ACROBiosystems AAVA005H96TESTS
Encompass_Preferred
AAV5 Titration ELISA Kit(Fast) is based on ELISA sandwich method and designed to detect and quantitative determine of AAV5 capsid titration in AAV gene therapy product preparation processing. It contains AAV5 empty capsids and a pair of antibodies against AAV5. Results are obtained by four parameter logistic curve that were parallel to the standard curves obtained. The verification results indicate that this kit can be used for the quantitative determination of AAV5 capsid titration.
Catalog No. 50-253-5531