Learn More
Teknova AAV Affinity Storage Buffer, 500mL, Sterile.
SDP
Catalog No. 502387466
Encompass
39mM Sodium Phosphate Monobasic, 61mM Sodium Phosphate Dibasic, 20% Ethanol, adjusted to pH 7.0
Catalog No. 50-238-7466 Supplier Teknova Supplier No. 20761
39mM Sodium Phosphate Monobasic, 61mM Sodium Phosphate Dibasic, 20% Ethanol, adjusted to pH 7.0