Learn More
BIOHIPPO AAV-ChAT-fDIO-hM3D(Gq)-mCherry-WPREs
SDP
Catalog No. 5000280077
Encompass
Recombinant adeno associated virus expressing hM3D(Gq) fused to mCherry under the ChAT promoter with WPRE and hGH polyA elements. The Flp dependent configuration enables expression only after recombination, providing precise chemogenetic control of targeted neurons. Offered under the Biohippo AAVX Series with multiple serotypes available including AAV2/1, AAV2/2, AAV2/5, AAV2/6, AAV2/6m, AAV2/6.2FF, AAV2/7m8, AAV2/8, AAV2/9, AAV2/DJ, AAV2/DJ8, AAV2/Retro, AAV2/PHP.eB, AAV2/PHP.B, AAV2/rh10, AAV2/PHP.S, AAV2/anc80, AAV2/B10, AAV2/olig001, AAV2/BI30, AAV2/BR1, AAV2/MG1.2, AAV1/2, AAV2/MaCPNS1, and AAV2/MaCPNS2. Regular titer is greater than or equal to 2.00E+12 vg per mL, with high titer greater than or equal to 1.00E+13 vg per mL available upon request at no additional cost. Customers may specify preferred serotype and titer when placing orders. The Cre dependent configuration enables expression only after recombination, providing precise chemogenetic control.
Catalog No. 50-002-80077 Supplier BIOHIPPO Supplier No. BHV12401561
Recombinant adeno associated virus expressing hM3D(Gq) fused to mCherry under the ChAT promoter with WPRE and hGH polyA elements. The Flp dependent configuration enables expression only after recombination, providing precise chemogenetic control of targeted neurons. Offered under the Biohippo AAVX Series with multiple serotypes available including AAV2/1, AAV2/2, AAV2/5, AAV2/6, AAV2/6m, AAV2/6.2FF, AAV2/7m8, AAV2/8, AAV2/9, AAV2/DJ, AAV2/DJ8, AAV2/Retro, AAV2/PHP.eB, AAV2/PHP.B, AAV2/rh10, AAV2/PHP.S, AAV2/anc80, AAV2/B10, AAV2/olig001, AAV2/BI30, AAV2/BR1, AAV2/MG1.2, AAV1/2, AAV2/MaCPNS1, and AAV2/MaCPNS2. Regular titer is greater than or equal to 2.00E+12 vg per mL, with high titer greater than or equal to 1.00E+13 vg per mL available upon request at no additional cost. Customers may specify preferred serotype and titer when placing orders. The Cre dependent configuration enables expression only after recombination, providing precise chemogenetic control.