Learn More
Cell Biolabs Inc AAV-DJ Helper Free Packaging System
Supplier:  Cell Biolabs Inc VPK400DJ
Encompass_Preferred
AAV-DJ is a hybrid AAV serotype developed from the recombination of 8 different native AAV serotypes. AAV-DJ viruses can infect a broad range of target cell types with high transduction efficieny. Cell Biolabs' AAV-DJ Helper Free Packaging System uses a Helper plasmid to eliminate the need for a helper virus in the packaging process. The system contains two packaging vectors (pHelper and a Rep-Cap vector) and a GFP control vector that can be used to make a GFP reporter virus. The system does not include a cloning vector, but may be used with the pAAV expression vector of your choice. FOR SALE TO ACADEMIC, GOVERNMENT AND NON-PROFIT INSTITUTIONS ONLY.
Catalog No. 50-673-805