Learn More
Agilent Technologies AAV HELPER-FREE SYSTEM
Catalog No. 50125418
Encompass
AAV Helper-Free System. Stable gene expression, broad host range, infects both dividing and non-dividing cells, up to 100% transduction efficiency, lower toxicity vs. non-viral methods of gene delivery
Catalog No. 50-125-418 Supplier Agilent Technologies Supplier No. 240071
AAV Helper-Free System. Stable gene expression, broad host range, infects both dividing and non-dividing cells, up to 100% transduction efficiency, lower toxicity vs. non-viral methods of gene delivery