Learn More
BIOHIPPO AAV-hSyn-EGFP-HA-WPRE-hGH-polyA
SDP
Catalog No. 5000279396
Encompass
Recombinant adeno associated virus expressing EGFP HA under the hSyn promoter with WPRE and hGH polyA elements. Offered under the Biohippo AAVX Series with multiple serotypes available including AAV2/1, AAV2/2, AAV2/5, AAV2/6, AAV2/6m, AAV2/6.2FF, AAV2/7m8, AAV2/8, AAV2/9, AAV2/DJ, AAV2/DJ8, AAV2/Retro, AAV2/PHP.eB, AAV2/PHP.B, AAV2/rh10, AAV2/PHP.S, AAV2/anc80, AAV2/B10, AAV2/olig001, AAV2/BI30, AAV2/BR1, AAV2/MG1.2, AAV1/2, AAV2/MaCPNS1, and AAV2/MaCPNS2. Regular titer is greater than or equal to 2.00E+12 vg per mL, with high titer greater than or equal to 1.00E+13 vg per mL available upon request at no additional cost. Customers may specify preferred serotype and titer when placing orders.
Catalog No. 50-002-79396 Supplier BIOHIPPO Supplier No. BHV12401032
Recombinant adeno associated virus expressing EGFP HA under the hSyn promoter with WPRE and hGH polyA elements. Offered under the Biohippo AAVX Series with multiple serotypes available including AAV2/1, AAV2/2, AAV2/5, AAV2/6, AAV2/6m, AAV2/6.2FF, AAV2/7m8, AAV2/8, AAV2/9, AAV2/DJ, AAV2/DJ8, AAV2/Retro, AAV2/PHP.eB, AAV2/PHP.B, AAV2/rh10, AAV2/PHP.S, AAV2/anc80, AAV2/B10, AAV2/olig001, AAV2/BI30, AAV2/BR1, AAV2/MG1.2, AAV1/2, AAV2/MaCPNS1, and AAV2/MaCPNS2. Regular titer is greater than or equal to 2.00E+12 vg per mL, with high titer greater than or equal to 1.00E+13 vg per mL available upon request at no additional cost. Customers may specify preferred serotype and titer when placing orders.