Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ AAV-MAX Control Plasmids Kit

Catalog No. A47672
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A47672 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A47672 Supplier Gibco™ Supplier No. A47672
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Gibco™ AAV-MAX Control Plasmids Kit consists of an optimized mix of three plasmids needed to produce adeno-associated virus serotype 2 (AAV2) expressing GFP: pAAV-CMV-emGFP, pAAV-Rep2Cap2, and pHelper.

The Gibco AAV-MAX Control Plasmids Kit consists of an optimized mix of three plasmids needed to produce adeno-associated virus serotype 2 (AAV2) expressing GFP: pAAV-CMV-emGFP, pAAV-Rep2Cap2, and pHelper. These three plasmids need to be used with a HEK293 cell line that stably expresses the adenovirus E1 gene. The kit contains an optimized mix of the three plasmids, as well as a separate tube of each of the three plasmids, which enables the user to utilize them in standalone manner and change the ratios.

• Saves process development time with optimized plasmid ratios
• Serves as a positive control for AAV2-GFP production with the AAV-MAX Helper-Free AAV Production system (Cat. No. A51217)

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
Product Type Plasmid
Product Line AAV-MAX
Quantity 1 Kit
Shipping Condition Ice
Format Kit
Regulatory Status RUO
Content And Storage • 1 tube containing mixture of all three plasmids, 1.5 mL
• 1 tube of each of the three plasmids alone, 0.75 mL

Store at –20°C.

Delivery Type Transfection
Expression System Mammalian

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.