Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ AAV-MAX Helper-Free AAV Production System Kit, without lysis buffer

Catalog No. A54824
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A54824 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A54824 Supplier Gibco™ Supplier No. A54824
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Gibco™ AAV-MAX Helper-Free AAV Production System is a scalable and high-titer platform for suspension AAV production. It is based on a high-density suspension culture of clonal 293F-derived Viral Production Cells 2.0 in Viral Production Medium.

  • A scalable and high-titer platform for suspension AAV production
  • Based on a high-density suspension culture of clonal 293F-derived Viral Production Cells 2.0 in Viral Production Medium
  • Optimal viral titers are mediated by our advanced lipid nanoparticle transfection reagent in combination with a novel production enhancer
  • The components are designed to work synergistically, resulting in maximal titers and eliminating the need to optimize reagents and protocols
  • Note, this version of the kit does not contain the AAV-MAX Lysis Buffer
  • To purchase the kit including the AAV-MAX Lysis Buffer, please see A51217
  • All components are also available for purchase separately

Features of the AAV-MAX Helper-Free AAV Production System include:

  • Components work synergistically to produce high AAV titers greater than 5x1010 vg/mL (unpurified)
  • Suspension-based system has scalable protocols from shake flask to bioreactor scale
  • Simplified workflow—streamlined protocol using helper virus-free triple transfection
  • Clonal, 293F-derived, high-producer cell line
  • Robust transfection protocols are adaptable to large scale transfection

Components of the AAV-MAX System Kit include:

  • 1 vial of Viral Production Cells 2.0
  • 1 L of Viral Production Medium
  • 1 AAV-MAX Transfection Kit for 1 L of production
  • 100 mL Viral-Plex Complexation Buffer
  • 100 mL GlutaMAX™ Supplement
TRUSTED_SUSTAINABILITY

Specifications

Cell Line Viral Production Cells 2.0
Cell Type Kidney (Embryonic)
Product Type AAV Production System
Sterility Sterile-filtered
For Use With (Application) AAV production
Product Line AAV-MAX
Quantity 1 Kit
Content And Storage • 1 vial Viral Production Cells 2.0
• 1 L Viral Production Medium
• 1 AAV-MAX Transfection kit (for 1 L production)
• 100 mL Viral-Plex Complexation Buffer
• 100 mL GlutaMAX

Culture Type Suspension Cell Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.