Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ AAV-MAX pHelper Vector

Catalog No. A47677
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A47677 1 Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A47677 Supplier Gibco™ Supplier No. A47677
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Gibco™ AAV-MAX Control Plasmids can be used to produce adeno-associated virus serotype 2 (AAV2) expressing GFP.

The Gibco AAV-MAX Control Plasmids can be used to produce adeno-associated virus serotype 2 (AAV2) expressing GFP. The three plasmids include: pAAV-CMV-emGFP, a pAAV-Rep2Cap2, and pHelper. These three plasmids need to be used with a HEK293 cell line that stably expresses the adenovirus E1 gene. The pAAV-CMV-emGFP is a transfer plasmid, the pAAV-Rep2Cap2 is a Rep/Cap plasmid, and the pHelper is a helper plasmid.

• Standalone plasmids provide flexibility to optimize plasmid ratios
• When used together, serve as a positive control for AAV2-GFP production with the AAV-MAX Helper-Free AAV Production system (Cat. No. A51217)

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
Product Type Plasmid
Sterility Non-sterile
Product Line AAV-MAX
Quantity 1 Tube
Shipping Condition Ice
Regulatory Status RUO
Content And Storage • 1 x 0.75 mL tube, 1 mg/mL

Store at -20°C.

Delivery Type Transfection
Expression System Mammalian

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.