Learn More
Norgen Biotek Corp AAV Purification Mini Kit
Catalog No. NC1752338
Encompass_Preferred
Recombinant adeno-associated virus (AAV) vectors are highly promising tools for both in vitro and in vivo gene transfer. Norgen’s AAV Purification Mini Kit provides a fast and simple procedure for concentrating and purifying AAV vectors from cell lysate and cell culture media. Purification is based on spin column chromatography using Norgen’s proprietary resin as an ion exchanger. Contaminating cellular debris is largely removed from the sample via a precipitation and centrifugation step, while contaminating DNA and RNA is reduced using enzymatic digestion. AAV vector purified in this manner is highly active for use in in vitro transduction experiments, and is eluted into a small volume (200 µL).
Catalog No. NC1752338 Supplier Norgen Biotek Corp Supplier No. 63200
Recombinant adeno-associated virus (AAV) vectors are highly promising tools for both in vitro and in vivo gene transfer. Norgen’s AAV Purification Mini Kit provides a fast and simple procedure for concentrating and purifying AAV vectors from cell lysate and cell culture media. Purification is based on spin column chromatography using Norgen’s proprietary resin as an ion exchanger. Contaminating cellular debris is largely removed from the sample via a precipitation and centrifugation step, while contaminating DNA and RNA is reduced using enzymatic digestion. AAV vector purified in this manner is highly active for use in in vitro transduction experiments, and is eluted into a small volume (200 µL).