Learn More
Cell Biolabs Inc ViraBind™ AAV Purification Kit, 10 preps
Catalog No. 50673754
Encompass_Preferred
Traditional ultracentrifugation procedures used to purify AAV preps are tedious, time consuming, and only partially purify the virus. Cell Biolabs' ViraBind™ AAV Purification Kit produces higher viral yield with higher purity than ultracentrifugation in a fraction of the time. Each kit is sufficient for 10 preps, and each prep can purify supernatant from two 10cm dishes or one 15cm dish.
Catalog No. 50-673-754 Supplier Cell Biolabs Inc Supplier No. VPK140
Traditional ultracentrifugation procedures used to purify AAV preps are tedious, time consuming, and only partially purify the virus. Cell Biolabs' ViraBind™ AAV Purification Kit produces higher viral yield with higher purity than ultracentrifugation in a fraction of the time. Each kit is sufficient for 10 preps, and each prep can purify supernatant from two 10cm dishes or one 15cm dish.