Learn More
Cell Biolabs Inc AAV Rep ELISA Kit, 96 assays
Supplier:  Cell Biolabs Inc VPK5118
Encompass_Preferred
Cell Biolabs' AAV Rep ELISA Kit quantifies the total of all 4 Rep proteins (Rep40, Rep52, Rep68, and Rep78) in AAV preps regardless of serotype. The limit of detection is 30 pg/mL of Rep protein. Each kit contains sufficient reagents for 96 assays including standards and unknown samples.
Catalog No. NC1712851