Learn More
Cell Biolabs Inc ViraDuctin™ AAV Transduction Kit, 50 transductions
Catalog No. 501098812
Encompass_Preferred
AAV can infect both dividing and non-dividing cells and can be maintained in the human host cell, creating the potential for genome integration. Because AAV is a naturally defective virus, requiring provision of several factors in trans for productive infection, it is considered the safest viral vector to use. The AAV transduction process includes viral binding and entry, intracellular trafficking, nuclear transport, and viral second strand DNA synthesis. The viral second strand DNA synthesis has been shown to be the rate limiting step, which leads to inefficient transduction by AAV vectors. ViraDuctin™ AAV Transduction Kit is a proprietary formulation designed to increase the transduction efficiency of AAV vectors in a variety of cell types. This kit provides the following advantages: (1) Higher transduction efficiency in many cell types, (2) Easy to use, and (3) It is ideal for transduction of nonpermissive cells such as primary cells and stem cells. 50 transductions/kit.
Catalog No. 50-109-8812 Supplier Cell Biolabs Inc Supplier No. AAV201
AAV can infect both dividing and non-dividing cells and can be maintained in the human host cell, creating the potential for genome integration. Because AAV is a naturally defective virus, requiring provision of several factors in trans for productive infection, it is considered the safest viral vector to use. The AAV transduction process includes viral binding and entry, intracellular trafficking, nuclear transport, and viral second strand DNA synthesis. The viral second strand DNA synthesis has been shown to be the rate limiting step, which leads to inefficient transduction by AAV vectors. ViraDuctin™ AAV Transduction Kit is a proprietary formulation designed to increase the transduction efficiency of AAV vectors in a variety of cell types. This kit provides the following advantages: (1) Higher transduction efficiency in many cell types, (2) Easy to use, and (3) It is ideal for transduction of nonpermissive cells such as primary cells and stem cells. 50 transductions/kit.