Learn More
BIOHIPPO AAV-U6--shRNA(scramble)--CMV-EGFP-SV40-polyA
SDP
Catalog No. 5000278958
Encompass
Recombinant adeno associated virus expressing shRNA (scramble) fused to EGFP under the U6 CMV promoter with WPRE and hGH polyA elements. Offered under the Biohippo AAVX Series with multiple serotypes available including AAV2/1, AAV2/2, AAV2/5, AAV2/6, AAV2/6m, AAV2/6.2FF, AAV2/7m8, AAV2/8, AAV2/9, AAV2/DJ, AAV2/DJ8, AAV2/Retro, AAV2/PHP.eB, AAV2/PHP.B, AAV2/rh10, AAV2/PHP.S, AAV2/anc80, AAV2/B10, AAV2/olig001, AAV2/BI30, AAV2/BR1, AAV2/MG1.2, AAV1/2, AAV2/MaCPNS1, and AAV2/MaCPNS2. Regular titer is greater than or equal to 2.00E+12 vg per mL, with high titer greater than or equal to 1.00E+13 vg per mL available upon request at no additional cost. Customers may specify preferred serotype and titer when placing orders.
Catalog No. 50-002-78958 Supplier BIOHIPPO Supplier No. BHV12400508
Recombinant adeno associated virus expressing shRNA (scramble) fused to EGFP under the U6 CMV promoter with WPRE and hGH polyA elements. Offered under the Biohippo AAVX Series with multiple serotypes available including AAV2/1, AAV2/2, AAV2/5, AAV2/6, AAV2/6m, AAV2/6.2FF, AAV2/7m8, AAV2/8, AAV2/9, AAV2/DJ, AAV2/DJ8, AAV2/Retro, AAV2/PHP.eB, AAV2/PHP.B, AAV2/rh10, AAV2/PHP.S, AAV2/anc80, AAV2/B10, AAV2/olig001, AAV2/BI30, AAV2/BR1, AAV2/MG1.2, AAV1/2, AAV2/MaCPNS1, and AAV2/MaCPNS2. Regular titer is greater than or equal to 2.00E+12 vg per mL, with high titer greater than or equal to 1.00E+13 vg per mL available upon request at no additional cost. Customers may specify preferred serotype and titer when placing orders.