Learn More
American Research Products Inc AAV1 ELISA Control | 1 vial (0.6E+09 – 1.0E+09 capsids)
Supplier:  American Research Products Inc PRAAV1C
Encompass
The AAV1 ELISA Control consists of fully assembled, empty AAV1 capsids. It represents the serotype-specific conformational epitopes and is therefore a reliable positive control in detection and quantification assays based on AAV antibodies detecting the conformational epitopes, such as ELISA.
Catalog No. NC1805760