Learn More
American Research Products Inc AAV1 Empty Capsids, > 95% Purity, > 5.0E+12 Capsids/ml, 100 µl
Catalog No. NC2096923
Encompass
AAV1 Empty Capsids are non-recombinant adeno-associated virus serotype 1 capsids produced in HEK293T cells, offering unmatched purity (> 95%) and high concentration (> 5.0E+12 capsids/ml). Ideal for various immunological applications like ELISA, dot blot, and Western blot, they feature a buffer formulation compatible with diverse experimental setups. Stored at -80°C for long-term use, they avoid the use of stabilizing proteins or dyes, making them a reliable choice for researchers.
This item is not returnable. View return policy
Catalog No. NC2096923 Supplier American Research Products Inc Supplier No. 66V010
AAV1 Empty Capsids are non-recombinant adeno-associated virus serotype 1 capsids produced in HEK293T cells, offering unmatched purity (> 95%) and high concentration (> 5.0E+12 capsids/ml). Ideal for various immunological applications like ELISA, dot blot, and Western blot, they feature a buffer formulation compatible with diverse experimental setups. Stored at -80°C for long-term use, they avoid the use of stabilizing proteins or dyes, making them a reliable choice for researchers.