Learn More
American Research Products Inc AAV-1 Titration ELISA | 96 tests
Supplier:  American Research Products Inc PRAAV1
Encompass
Enzyme Immunoassay for the Quantitative Determination of AAV Serotype 1 Particles in Cell Culture Supernatants and Purified Virus Preparations.
Catalog No. NC1805759