Learn More
American Research Products Inc AAV10 Titration ELISA Kit, Highly Sensitive Sandwhich ELISA, Monoclonal Antibody Based, 96 tests
Supplier:  American Research Products Inc PRAAV10
Encompass
The AAVrh10 Titration ELISA Kit offers a highly sensitive and reproducible method for quantifying AAVrh10 virions and assembled empty capsids in research. With a monoclonal capture antibody targeting specific AAVrh10 capsid epitopes, it ensures precise measurement. Easy to use and efficient with a colorimetric detection method, this kit is ideal for various AAV applications. Suitable for research use only, it streamlines workflows and saves time compared to traditional methods.
Catalog No. NC2079592