Learn More
American Research Products Inc AAV2 Titration ELISA 2.0R, Sandwich ELISA, Detects AAV2 Capsids, 96 Tests
Supplier:  American Research Products Inc PRAAV2R
Encompass
The AAV2 Titration ELISA 2.0R is an advanced sandwich ELISA kit designed for the precise quantitation of AAV2 capsids in research. This assay offers superior reliability compared to traditional detection methods, thanks to its recombinant antibody targeting specific epitopes, ensuring high sensitivity and minimal variability. With a capacity of 96 tests, it provides ease of use and robust performance for researchers focused on viral quantitation.
Catalog No. NC1941877