Learn More
XpressBio AAV2 ELISA Assay, 96 wells
SDP
Supplier:  XpressBio XBAAV21000
Encompass_Preferred
The AAV2 ELISA Assay can be used to measure titrations for AAV2; including AAV2 wildtype virions, assembled/intact empty AAV2 capsids, and recombinant virions.
Catalog No. NC2462574