Learn More
Genscript Corporation AAV2 Titer Capsid ELISA Kit
Supplier:  Genscript Corporation L00942
Encompass
GenScript AAV2 Titer Capsid ELISA Kit (Cat. No. L00942) is a Sandwich ELISA Kit that is designed for quantitatively measuring the total capsid titer of adeno-associated virus serotype 2 (AAV2) in testing samples during viral vector production. The total capsid titer stands for the amount of all fully assembled viral capsids, including full and empty AAV2 capsids. This ELISA kit provides an easy, accurate, and reproducible method for AAV2 titration.
Catalog No. NC2229426