Learn More
SignaGen Laboratories AAV-GFP (AAV Serotype 5), 30 uL
Catalog No. NC1821236
Encompass
AAV5-GFP is a pre-packaged rAAV in serotype 5 (with capsid from AAV serotype 5 and 2xITR from AAV serotype 2) which over-expresses EGFP under CMV promoter. Ready to use format.
Catalog No. NC1821236 Supplier SignaGen Laboratories Supplier No. SL100819
AAV5-GFP is a pre-packaged rAAV in serotype 5 (with capsid from AAV serotype 5 and 2xITR from AAV serotype 2) which over-expresses EGFP under CMV promoter. Ready to use format.