Learn More
American Research Products Inc AAV5 Titration ELISA Kit, Quantitative AAV5 Capsid Detection, 96 Tests
Supplier:  American Research Products Inc PRAAV5
Encompass
The AAV5 Titration ELISA Kit is a fast and reliable tool for quantifying AAV5 capsids, enabling effective validation of AAV gene therapy safety and efficacy. With a streamlined process yielding results in under 4.5 hours, this kit stands out for its accurate assessment of both full and empty virus particles, utilizing a specific monoclonal antibody for detection. Ideal for research purposes, it facilitates important advancements in gene therapy research.
Catalog No. NC1572423