Learn More
American Research Products Inc AAV8 Empty Capsids, Highly Purified, Ideal For Immunoassays, 100 mL
Catalog No. NC2105298
Encompass
AAV8 Empty Capsids are highly purified, fully assembled empty capsids suitable for a variety of scientific applications. They are produced in HEK293T cells and are quality-controlled for high purity and consistent titer. This product is ideal for serving as a positive control in assays like dot blot, ELISA, and western blot, making it a dependable resource for research. With over 95% purity and an endotoxin level of less than 1.0 EU/ml, it meets high standards for laboratory use.
Catalog No. NC2105298 Supplier American Research Products Inc Supplier No. 66V080
AAV8 Empty Capsids are highly purified, fully assembled empty capsids suitable for a variety of scientific applications. They are produced in HEK293T cells and are quality-controlled for high purity and consistent titer. This product is ideal for serving as a positive control in assays like dot blot, ELISA, and western blot, making it a dependable resource for research. With over 95% purity and an endotoxin level of less than 1.0 EU/ml, it meets high standards for laboratory use.