Learn More
American Research Products Inc AAV8 XPRESS ELISA, Rapid Assay, High Sensitivity, Research Use Only, 96 Tests
Supplier:  American Research Products Inc PRAAV8XP
Encompass
The AAV8 XPRESS ELISA is a rapid, sensitive sandwich ELISA designed for the precise quantification of AAV8 capsid titers in various biological fluids. It significantly reduces assay time to under 2 hours without compromising accuracy, making it a reliable choice for researchers. The assay utilizes a specific monoclonal antibody to ensure the quantification of intact AAV8 virions and provides consistent results akin to traditional methods.
Catalog No. NC1955870