Learn More
American Research Products Inc AAV9 Empty Capsids, > 95% Purity, Verified by SDS-PAGE, 100 µl
Catalog No. NC2096927
Encompass
AAV9 Empty Capsids offer high purity, produced in HEK293T cells, and designed for sensitive assays like dot blot, western blot, and ELISA. Supplied by AMERICAN RESEARCH PRODUCTS INC, these liquid capsids are ideal for gene therapy research, distinguishing themselves with a titer of > 5.0E+12 viral particles/ml and a stable formulation devoid of stabilizers. They facilitate accurate assay validation with a short-term storage option of 2 weeks at 2-8°C, ensuring reliable performance in various scientific applications.
This item is not returnable. View return policy
Catalog No. NC2096927 Supplier American Research Products Inc Supplier No. 66V090
AAV9 Empty Capsids offer high purity, produced in HEK293T cells, and designed for sensitive assays like dot blot, western blot, and ELISA. Supplied by AMERICAN RESEARCH PRODUCTS INC, these liquid capsids are ideal for gene therapy research, distinguishing themselves with a titer of > 5.0E+12 viral particles/ml and a stable formulation devoid of stabilizers. They facilitate accurate assay validation with a short-term storage option of 2 weeks at 2-8°C, ensuring reliable performance in various scientific applications.