Learn More
American Research Products Inc AAV9 Xpress ELISA | 96 tests
Supplier:  American Research Products Inc PRAAV9XP
Encompass
Fast, easy and reliable AAV9 titer determination with PROGEN's AAV9 Xpress ELISA: Assay time was reduced to less than 2 hours by adjustment of the kit components and the protocol of our standard AAV9 Titration ELISA. Seamless and risk-free transition from our standard AAV9 Titration ELISAs (PRAAV9) to the Xpress version since the composition of the kit’s core characteristics are not changed. Same accuracy and reproducibility as the standard AAV9 Titration ELISAs.
Catalog No. NC1872868
 Restricted Item