Learn More
Takara Bio AAVpro 293T Cell Line | 1mL
SDP
Catalog No. NC1415837
Encompass
The AAVpro 293T Cell Line is a subclone of the transformed human embryonic kidney cell line HEK 293, which is highly transfectable and supports high levels of viral protein expression. The cell line also constitutively expresses the simian virus 40 (SV40) large T antigen. When combined with the AAVpro Helper Free System (Cat. No. 6230), these cells can produce high titers of recombinant adeno-associated virus (AAV).
Catalog No. NC1415837 Supplier Takara Bio Supplier No. 632273
The AAVpro 293T Cell Line is a subclone of the transformed human embryonic kidney cell line HEK 293, which is highly transfectable and supports high levels of viral protein expression. The cell line also constitutively expresses the simian virus 40 (SV40) large T antigen. When combined with the AAVpro Helper Free System (Cat. No. 6230), these cells can produce high titers of recombinant adeno-associated virus (AAV).