Learn More
Takara Bio AAVPRO PURIFICATION KIT MIDI
SDP
Catalog No. NC1847372
Encompass
AAVpro Purification Kit Midi (All Serotypes),4 Preps.The AAVpro Purification Kit Midi (All Serotypes) is a four-prep kit that can be used for purifying AAV of any serotype without the need for ultracentrifugation. The purification yields highly pure AAV particles that can be used directly for in vivo transduction or transduction of cultured cells. Cryonase Cold-Active Nuclease is included. This product allows for purification of four virus preps, each from a culture size of 1 x T225 flask.
Catalog No. NC1847372 Supplier Takara Bio Supplier No. 6675
AAVpro Purification Kit Midi (All Serotypes),4 Preps.The AAVpro Purification Kit Midi (All Serotypes) is a four-prep kit that can be used for purifying AAV of any serotype without the need for ultracentrifugation. The purification yields highly pure AAV particles that can be used directly for in vivo transduction or transduction of cultured cells. Cryonase Cold-Active Nuclease is included. This product allows for purification of four virus preps, each from a culture size of 1 x T225 flask.