Learn More
System Biosciences LLC AAVS1 Safe Harbor Site Targeting Knock-in HR Donor Vector 2.0 (AAVS1-SA-puro-EF1α-MCS)AAVS1 Safe Harbor Targeting Knock-in HR Donor Vector 2.0 - Knock-in your gene-of-interest driven by the EF1α promoter (AAVS1-SA-puro-EF1α-MCS) = 10 ug
SDP
Catalog No. NC1614255
Encompass_Preferred
Easily knock-in any gene into the AAVS1 Safe Harbor Site—your gene-of-interest is driven by the constitutive EF1α promoter. When you want to take advantage of the robust and reliable expression that’s possible from the AAVS1 Safe Harbor Site, SBI offers a range of second generation (2.0) AAVS1 Safe Harbor Site Targeting HR Donor Vectors that are designed minimize off-target integration. The AAVS1-Targeting Knock-in HR Donor Vector 2.0 AAVS1-SA-puro-EF1α-MCS contains a multiple cloning site (MCS) downstream of the EF1α promoter, enabling constitutive expression of your gene-of-interest from the AAVS1 locus.
Catalog No. NC1614255 Supplier System Biosciences LLC Supplier No. GE622A1
Easily knock-in any gene into the AAVS1 Safe Harbor Site—your gene-of-interest is driven by the constitutive EF1α promoter. When you want to take advantage of the robust and reliable expression that’s possible from the AAVS1 Safe Harbor Site, SBI offers a range of second generation (2.0) AAVS1 Safe Harbor Site Targeting HR Donor Vectors that are designed minimize off-target integration. The AAVS1-Targeting Knock-in HR Donor Vector 2.0 AAVS1-SA-puro-EF1α-MCS contains a multiple cloning site (MCS) downstream of the EF1α promoter, enabling constitutive expression of your gene-of-interest from the AAVS1 locus.