Learn More
Cayman Chemical AB-005 azepane isomer, Purity: ≥98%, CAS: 1445751-74-5, 10 mg
SDP
Catalog No. 502951987
Encompass_Preferred
A variant of AB-005 in which the methylpiperidine group has been replaced with methylazepane; intended for forensic and research applications
Catalog No. 50-295-1987 Supplier Cayman Chemical Supplier No. 1187710 MG
A variant of AB-005 in which the methylpiperidine group has been replaced with methylazepane; intended for forensic and research applications