Learn More
Dojindo Molecular Technologies Inc Ab-10 Rapid Biotin Labeling Kit (3 samples) is used to prepare biotin-labeled antibodies from 10 µg protein samples, just mix antibody and reagent, recover conjugates within 30 min.
SDP
Supplier:  Dojindo Molecular Technologies Inc LK3710
Encompass_Preferred
Ab-10 Rapid Biotin Labeling Kit is used for the preparation of biotin-labeled IgG for enzyme immunoassay (EIA). This kit enables rapid (in less than 30 min) and easy binding of Biotin to 10 μg antibody. This kit contains all the necessary reagents for antibody labeling.Reactive Biotin (a component of the kit) has a succinimidyl ester group that can easily make a covalent bond with an amino group of the target antibody without any activation process. The procedure is very simple: just mix antibody and labeling agent. Dojindo offers various protein labeling reagents. (https://www.dojindo.com/labeling-chemistry)
Catalog No. NC1960977