Learn More
Dojindo Molecular Technologies Inc Ab-10 Rapid Fluorescein Labeling Kit (3 samples) is used to prepare fluorescein-labeled antibody from 10μg protein samples, just mix antibody and reagent; recover conjugates within 30 min.
SDP
Catalog No. NC1960981
Encompass_Preferred
Ab-10 Rapid Fluorescein Labeling Kit is used mainly for the preparation of fluorescein-labeledIgG for enzyme immunoassay (EIA) and for the preparation of peroxidase-labeled antigen for competitive EIA. This kit can label fluorescein to 10 µg antibody in less than 30 minutes. NH2 reactive fluorescein, a component of the kit has a succinimidyl ester group, that can easily make a covalent bond with an amino group of the target antibody without any activation process. The kit contains all the necessary reagents for antibody labeling. The procedure is very simple: just mix antibody and labeling agent. Dojindo has various protein labeling reagents. (https://www.dojindo.com/labeling-chemistry/)
Catalog No. NC1960981 Supplier Dojindo Molecular Technologies Inc Supplier No. LK3210
Ab-10 Rapid Fluorescein Labeling Kit is used mainly for the preparation of fluorescein-labeledIgG for enzyme immunoassay (EIA) and for the preparation of peroxidase-labeled antigen for competitive EIA. This kit can label fluorescein to 10 µg antibody in less than 30 minutes. NH2 reactive fluorescein, a component of the kit has a succinimidyl ester group, that can easily make a covalent bond with an amino group of the target antibody without any activation process. The kit contains all the necessary reagents for antibody labeling. The procedure is very simple: just mix antibody and labeling agent. Dojindo has various protein labeling reagents. (https://www.dojindo.com/labeling-chemistry/)