Learn More
Dojindo Molecular Technologies Inc Ab-10 Rapid HiLyte Fluor 555 Labeling Kit (3 samples) is used to prepare HiLyte Fluor 555-labeled antibody from 10 µg protein samples, just mix antibody and reagent, recover conjugates within 30 min.
SDP
Catalog No. NC1960975
Encompass_Preferred
Ab-10 Rapid HiLyte FluorTM 555 Labeling Kit is used to prepare of red fluorescence-labeled proteins, such as IgG, for immunostaining and to prepare labeled cellular proteins for tracing. This kit enables rapid (in less than 30 min) and easy binding of HiLyte Fluor 555 to 10 µg of antibody. This kit contains all the necessary reagents for antibody labeling.Reactive HiLyte Fluor 555 (a component of the kit) has a succinimidyl ester group that can easily make a covalent bond with an amino group of the target antibody without any activation process. The procedure is very simple: just mix antibody and labeling agent. *HiLyte Fluor is a trademark of AnaSpec, Inc. Dojindo offers various protein labeling reagents. (https://www.dojindo.com/labeling-chemistry/
Catalog No. NC1960975 Supplier Dojindo Molecular Technologies Inc Supplier No. LK3510
Ab-10 Rapid HiLyte FluorTM 555 Labeling Kit is used to prepare of red fluorescence-labeled proteins, such as IgG, for immunostaining and to prepare labeled cellular proteins for tracing. This kit enables rapid (in less than 30 min) and easy binding of HiLyte Fluor 555 to 10 µg of antibody. This kit contains all the necessary reagents for antibody labeling.Reactive HiLyte Fluor 555 (a component of the kit) has a succinimidyl ester group that can easily make a covalent bond with an amino group of the target antibody without any activation process. The procedure is very simple: just mix antibody and labeling agent. *HiLyte Fluor is a trademark of AnaSpec, Inc. Dojindo offers various protein labeling reagents. (https://www.dojindo.com/labeling-chemistry/