Learn More
MSC Olfa Replacement Blades, 9mm, 13-Segment, Carbon Tool Steel, 10 Blades
Catalog No. 501111054
Encompass
These 9mm snap-off blades are crafted from high-quality carbon tool steel, designed for superior sharpness and excellent edge retention. They are suitable for general crafts and cutting various materials such as packaging, vinyl, wallpaper, and foam board.
  • Snap-off blade design
  • Made from high-quality carbon tool steel
  • Double-honed blade provides superior initial sharpness and edge retention
  • Exact 59-degree edge angle optimizes cutting power and minimizes blade breakage
Catalog No. 50-111-1054 Supplier MSC Supplier No. 32993776
These 9mm snap-off blades are crafted from high-quality carbon tool steel, designed for superior sharpness and excellent edge retention. They are suitable for general crafts and cutting various materials such as packaging, vinyl, wallpaper, and foam board.
  • Snap-off blade design
  • Made from high-quality carbon tool steel
  • Double-honed blade provides superior initial sharpness and edge retention
  • Exact 59-degree edge angle optimizes cutting power and minimizes blade breakage