Learn More
Medchemexpress LLC HY-125286 5mg , AB-680 CAS:2105904-82-1 Purity:>98%
SDP
Catalog No. 502259095
Encompass_Preferred
HY-125286 5mg AB-680 CAS: 2105904-82-1 AB-680 is a highly potent, reversible and selective inhibitor of CD73 (an ecto-nucleotidase), with a Ki of 4.9 pM for hCD73, displays >10,000-fold selectivity over related ecto-nucleotidases CD39. Anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9095 Supplier Medchemexpress LLC Supplier No. HY1252865MG
HY-125286 5mg AB-680 CAS: 2105904-82-1 AB-680 is a highly potent, reversible and selective inhibitor of CD73 (an ecto-nucleotidase), with a Ki of 4.9 pM for hCD73, displays >10,000-fold selectivity over related ecto-nucleotidases CD39. Anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.