Learn More
Full Moon Biosystems AB ARRAY ASSAY KIT 2REACTIONS
SDP
Catalog No. NC9689220
Encompass
2-reaction Antibody Array Assay Kit can be used to process 2 array slides Optimized to work with all Full Moon BioSystems' antibody microarrays and maximize protein yield, labeling efficiency Provides major reagents required for lysate preparation, protein labeling, coupling and detection Efficient, reliable and easy to use Kit Components: Biotin Reagent Blocking Reagent Coupling Chamber Coupling Reagent Detection Buffer Extraction Buffer Dry Milk Powder Labeling Buffer Spin Columns Stop Reagent 10X Wash Buffer
Catalog No. NC9689220 Supplier Full Moon Biosystems Supplier No. KAS02
2-reaction Antibody Array Assay Kit can be used to process 2 array slides Optimized to work with all Full Moon BioSystems' antibody microarrays and maximize protein yield, labeling efficiency Provides major reagents required for lysate preparation, protein labeling, coupling and detection Efficient, reliable and easy to use Kit Components: Biotin Reagent Blocking Reagent Coupling Chamber Coupling Reagent Detection Buffer Extraction Buffer Dry Milk Powder Labeling Buffer Spin Columns Stop Reagent 10X Wash Buffer