Learn More
Sigma Aldrich Fine Chemicals Biosciences Ab Buffer Kit Cytiva 28-90
Catalog No. 502900368
Encompass
Ab Buffer Kit contains carefully prepared buffer concentrates for binding washing and elution of IgG according to recommended protocols. The kit also includes neutralizing buffer. Includes 1 x 50 ml Binding buffer 10x stock SOLUTIONon 1 x 15 ml Elution buffer 10x stock SOLUTIONon and 1 x 25 ml Neutralizing buffer.
Catalog No. 50-290-0368 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28903059
Ab Buffer Kit contains carefully prepared buffer concentrates for binding washing and elution of IgG according to recommended protocols. The kit also includes neutralizing buffer. Includes 1 x 50 ml Binding buffer 10x stock SOLUTIONon 1 x 15 ml Elution buffer 10x stock SOLUTIONon and 1 x 25 ml Neutralizing buffer.