Learn More
Cayman Chemical AB-CHMINACA metabolite M3A, Purity: ≥98% (mixture of diastereomers), CAS: 2131173-58-3, 1 mg
SDP
Catalog No. 502970241
Encompass_Preferred
An expected metabolite produced during metabolism of AB-CHMINACA in the liver; intended for forensic and research applications
Catalog No. 50-297-0241 Supplier Cayman Chemical Supplier No. 163911 MG
An expected metabolite produced during metabolism of AB-CHMINACA in the liver; intended for forensic and research applications