Learn More
Cayman Chemical AB-CHMINACA metabolite M4, Purity: ≥98%, CAS: 1271630-11-5, 1 mg
SDP
Catalog No. 502970527
Encompass_Preferred
An expected metabolite of AB-CHMINACA in which the carboxamide group has been replaced with a carboxyl group; intended for research and forensic applications
Catalog No. 50-297-0527 Supplier Cayman Chemical Supplier No. 163931 MG
An expected metabolite of AB-CHMINACA in which the carboxamide group has been replaced with a carboxyl group; intended for research and forensic applications